D-(-)-Leucinol 95%

Supplier: Ambeed
Warning

Synonyms: D-Leucinol, (R)-(-)-Leucinol, (R)-(-)-2-Amino-4-methylpentan-1-ol, (R)-2-Amino-4-methylpentan-1-ol

A119293-10G A119293-1G A119293-25G A119293-5G A119293-100G
76971-400EA 45.62 USD
76971-400 76971-402 76971-404 77571-148 77610-998
D-(-)-Leucinol 95%
D-(-)-Leucinol

Precaución: for Research Use Only

Formula: C₆H₁₅NO
MW: 117.19 g/mol
MDL Number: MFCD00004734
CAS Number: 53448-09-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR