Fmoc-D-His-OH 95%

Supplier: Ambeed
Warning

Synonyms: (R)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-(1H-imidazol-4-yl)propanoic acid

A579493-25G A579493-5G A579493-1G
76980-378EA 381.93 USD
76980-378 76980-380 77673-554
Fmoc-D-His-OH 95%
Fmoc-D-His-OH

Precaución: for Research Use Only

Formula: C₂₁H₁₉N₃O₄
MW: 377.4 g/mol
MDL Number: MFCD00237022
CAS Number: 157355-79-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR