2-Iodo-1,3-dimethoxybenzene 97%

Supplier: Ambeed
Warning

A253420-100G A253420-10G A253420-1G A253420-250MG A253420-25G A253420-5G
76976-720EA 313 USD
76976-720 76976-722 76976-724 76976-726 76976-728 76976-730
2-Iodo-1,3-dimethoxybenzene 97%
2-Iodo-1,3-dimethoxybenzene

Precaución: for Research Use Only

Formula: C₈H₉IO₂
MDL Number: MFCD05865214
CAS Number: 16932-44-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR