(1S,2R)-N1-(tert-Butoxycarbonyl)-1,2-cyclohexanediamine 96%

Supplier: Ambeed
Warning

Synonyms: (1S,2R)-N1-Boc-1,2-cyclohexanediamine, (1S,2R)-2-Amino-1-(tert-butoxycarbonylamino)cyclohexane, tert-Butyl ((1S,2R)-2-aminocyclohexyl)carbamate, (1S,2R)-2-Amino-1-(Boc-amino)cyclohexane

A140823-250MG A140823-5G A140823-1G
76972-606EA 53.05 USD
76972-606 76972-608 77611-282
(1S,2R)-N1-(tert-Butoxycarbonyl)-1,2-cyclohexanediamine 96%
(1S,2R)-N1-(tert-Butoxycarbonyl)-1,2-cyclohexanediamine

Precaución: for Research Use Only

Formula: C₁₁H₂₂N₂O₂
MW: 214.31 g/mol
MDL Number: MFCD09952105
CAS Number: 365996-30-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR