2-Iodophenol 98%
Supplier: Ambeed
|
Warning
|
A159770-10G
A159770-500G
A159770-5G
A159770-25G
A159770-100G
76973-492EA
10.61
USD
76973-492
76973-494
76973-496
77300-412
77567-328
2-Iodophenol 98%
2-Iodophenol
Precaución: For research use only.
Formula:
C₆H₅IO MW: 220.01 g/mol |
MDL Number:
MFCD00013963 CAS Number: 533-58-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...