6,7-Dihydro-5H-cyclopenta[b]pyridine 95%

Supplier: Ambeed
Warning

Synonyms: 5H-1-Pyrindine, 6,7-dihydro-, 2,3-Cyclopentenopyridine

A149316-100G A149316-10G A149316-25G A149316-5G
76973-040EA 51.99 USD
76973-040 76973-042 76973-044 76973-046
6,7-Dihydro-5H-cyclopenta[b]pyridine 95%
6,7-Dihydro-5H-cyclopenta[b]pyridine

Precaución: for Research Use Only

Formula: C₈H₉N
MW: 119.17 g/mol
MDL Number: MFCD00005933
CAS Number: 533-37-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR