Sesamol-(3,4-(methylenedioxy)phenol) 97%

Supplier: Ambeed
Warning

Synonyms: Sesamol, 3,4-Methylenedioxyphenol, Benzo[d][1,3]dioxol-5-ol

A569861-100G A569861-100MG A569861-10G A569861-25G A569861-500G A569861-5G
76980-306EA 55.17 USD
76980-306 76980-308 76980-310 76980-312 76980-314 76980-316
Sesamol-(3,4-(methylenedioxy)phenol) 97%
Sesamol-(3,4-(methylenedioxy)phenol)

Precaución: for Research Use Only

Formula: C₇H₆O₃
MW: 138.12 g/mol
MDL Number: MFCD00005827
CAS Number: 533-31-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR