Ethyl-5-amino-2,6-dichloropyrimidine-4-carboxylate 95%
Supplier: Ambeed
|
Warning
|
A122407-100MG
A122407-1G
A122407-250MG
A122407-5G
76971-620EA
13
USD
76971-620
76971-622
76971-624
76971-626
Ethyl-5-amino-2,6-dichloropyrimidine-4-carboxylate 95%
Ethyl-5-amino-2,6-dichloropyrimidine-4-carboxylate
Precaución: for Research Use Only
Formula:
C₇H₇Cl₂N₃O₂ |
MDL Number:
MFCD18452009 CAS Number: 54368-62-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...