N-(tert-Butoxycarbonyl)-L-prolinamide 97%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl (S)-2-Carbamoyl-1-pyrrolidinecarboxylate, (S)-2-Carbamoyl-1-pyrrolidinecarboxylic Acid tert-Butyl Ester, 1-Boc-L-prolinamide, N-Boc-L-prolinamide, Boc-Pro-NH2

A190688-100G A190688-10G A190688-1G A190688-25G A190688-500G A190688-5G
76974-848EA 102.91 USD
76974-848 76974-850 76974-852 76974-854 76974-856 76974-858
N-(tert-Butoxycarbonyl)-L-prolinamide 97%
N-(tert-Butoxycarbonyl)-L-prolinamide

Precaución: for Research Use Only

Formula: C₁₀H₁₈N₂O₃
MW: 214.27 g/mol
MDL Number: MFCD00190827
CAS Number: 35150-07-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR