3-Amino-4-chlorobenzonitrile 97%

Supplier: Ambeed
Warning

A273820-100G A273820-10G A273820-1G A273820-25G A273820-5G
76977-248EA 82 USD
76977-248 76977-250 76977-252 76977-254 76977-256
3-Amino-4-chlorobenzonitrile 97%
3-Amino-4-chlorobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₅ClN₂
MDL Number: MFCD00017089
CAS Number: 53312-79-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR