1-Chloro-2,7-naphthyridine 98%

Supplier: Ambeed
Warning

A791173-1G A791173-250MG
76981-614EA 78 USD
76981-614 77674-148
1-Chloro-2,7-naphthyridine 98%
1-Chloro-2,7-naphthyridine

Precaución: for Research Use Only

Formula: C₈H₅ClN₂
MDL Number: MFCD00234389
CAS Number: 69042-30-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR