5,6-Dihydroxy-2,3-dihydro-1H-inden-1-one 98%

Supplier: Ambeed
Warning

A319098-250MG A319098-1G
76978-040EA 8 USD
76978-040 77550-890
5,6-Dihydroxy-2,3-dihydro-1H-inden-1-one 98%
5,6-Dihydroxy-2,3-dihydro-1H-inden-1-one

Precaución: for Research Use Only

Formula: C₉H₈O₃
MW: 164.16 g/mol
MDL Number: MFCD12756505
CAS Number: 124702-80-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR