2-Fluoro-4-iodonitrobenzene 97%

Supplier: Ambeed
Warning

Synonyms: 2-Fluoro-4-iodo-1-nitrobenzene

A139722-100MG A139722-1G A139722-250MG A139722-5G
76972-534EA 23.34 USD
76972-534 76972-536 76972-538 77611-272
2-Fluoro-4-iodonitrobenzene 97%
2-Fluoro-4-iodonitrobenzene

Precaución: for Research Use Only

Formula: C₆H₃FINO₂
MW: 266.996 g/mol
MDL Number: MFCD18396989
CAS Number: 2996-31-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR