4-(Bromomethyl)-2,2-dimethyl-1,3-dioxolane 98%
Supplier: Ambeed
|
Danger
|
A761451-1G
A761451-250MG
A761451-5G
A761451-25G
76981-426EA
26
USD
76981-426
76981-428
77552-332
77882-654
4-(Bromomethyl)-2,2-dimethyl-1,3-dioxolane 98%
4-(Bromomethyl)-2,2-dimethyl-1,3-dioxolane
Precaución: for Research Use Only
Formula:
C₆H₁₁BrO₂ |
MDL Number:
MFCD14584455 CAS Number: 36236-76-7 UN: 1760 ADR: 8,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...