3-Bromo-6-chloropyridin-2(1H)-one 96%

Supplier: Ambeed
Warning

A230523-100MG A230523-1G A230523-5G A230523-250MG
76976-132EA 59 USD
76976-132 76976-134 76976-136 77672-046
3-Bromo-6-chloropyridin-2(1H)-one 96%
3-Bromo-6-chloropyridin-2(1H)-one

Precaución: for Research Use Only

Formula: C₅H₃BrClNO
MDL Number: MFCD22123755
CAS Number: 848423-85-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR