1-(Phenoxycarbonyl)pyrrolidine-2-carboxylic acid 97%

Supplier: Ambeed
Warning

A114578-1G A114578-250MG
76971-102EA 62.6 USD
76971-102 76971-104
1-(Phenoxycarbonyl)pyrrolidine-2-carboxylic acid 97%
1-(Phenoxycarbonyl)pyrrolidine-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₃NO₄
MW: 235.2400 g/mol
MDL Number: MFCD01045469
CAS Number: 1161602-22-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR