Trimethylamine-N-oxide dihydrate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Trimethylamine oxide dihydrate
A690734-1000G
A690734-100G
A690734-25G
A690734-500G
76981-036EA
128.37
USD
76981-036
76981-038
76981-040
76981-042
Trimethylamine-N-oxide dihydrate 98%
Trimethylamine N-oxide dihydrate
Precaución: for Research Use Only
Formula:
C₃H₉NO·2H₂O MW: 111.14 g/mol (75.11 anhy) |
MDL Number:
MFCD00149077 CAS Number: 62637-93-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...