5-Fluoro-4-methyl-2-nitroanisole 98%

Supplier: Ambeed
Warning

Synonyms: 1-Fluoro-5-methoxy-2-methyl-4-nitrobenzene

A385920-100MG A385920-1G A385920-250MG A385920-25G A385920-5G
76978-870EA 64.72 USD
76978-870 76978-872 76978-874 76978-876 76978-878
5-Fluoro-4-methyl-2-nitroanisole 98%
5-Fluoro-4-methyl-2-nitroanisole

Precaución: for Research Use Only

Formula: C₈H₈FNO₃
MW: 185.152 g/mol
MDL Number: MFCD15527524
CAS Number: 314298-13-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR