N-(3-Trifluoromethylphenyl)anthranilic acid 98%

Supplier: Ambeed
Danger

A416271-100MG A416271-250MG A416271-1MG A416271-25G A416271-5G
76979-218EA 7 USD
76979-218 76979-220 77673-022 77882-091 77882-092
N-(3-Trifluoromethylphenyl)anthranilic acid 98%
N-(3-Trifluoromethylphenyl)anthranilic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₀F₃NO₂
MDL Number: MFCD00002422
CAS Number: 530-78-9
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR