2-Nitro-p-phenylenediamine 97%

Supplier: Ambeed
Warning

Synonyms: 2-Nitro-1,4-phenylenediamine, 2-Nitrobenzene-1,4-diamine

A133306-100G A133306-25G
76972-216EA 26.53 USD
76972-216 77298-618
2-Nitro-p-phenylenediamine 97%
2-Nitro-p-phenylenediamine

Precaución: for Research Use Only

Formula: C₆H₇N₃O₂
MW: 153.14 g/mol
MDL Number: MFCD00007903
CAS Number: 5307-14-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR