Benzo[d][1,3]dioxol-4-ylboronic acid 97%
Supplier: Ambeed
|
Warning
|
A571569-100MG
A571569-1G
A571569-250MG
76980-340EA
36
USD
76980-340
76980-342
77673-536
Benzo[d][1,3]dioxol-4-ylboronic acid 97%
Benzo[d][1,3]dioxol-4-ylboronic acid
Precaución: for Research Use Only
Formula:
C₇H₇BO₄ |
MDL Number:
MFCD03095113 CAS Number: 361456-68-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...