5-Bromo-2,3-diphenyl-1H-indole 95%

Supplier: Ambeed
Warning

A255725-100MG A255725-1G A255725-250MG A255725-5G
76976-822EA 8 USD
76976-822 76976-824 76976-826 76976-828
5-Bromo-2,3-diphenyl-1H-indole 95%
5-Bromo-2,3-diphenyl-1H-indole

Precaución: for Research Use Only

Formula: C₂₀H₁₄BrN
MDL Number: MFCD20257791
CAS Number: 1259224-11-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR