3-Bromo-6,9-diphenylcarbazole 97%

Supplier: Ambeed
Warning

A171806-1G A171806-25G A171806-5G
76974-030EA 7 USD
76974-030 76974-032 76974-034
3-Bromo-6,9-diphenylcarbazole 97%
3-Bromo-6,9-diphenylcarbazole

Precaución: for Research Use Only

Formula: C₂₄H₁₆BrN
MDL Number: MFCD20257783
CAS Number: 1160294-85-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR