4-Chloro-6-methyl-5H-pyrrolo[3,2-d]pyrimidine 95%

Supplier: Ambeed
Warning

A717048-1G A717048-250MG A717048-5G
76981-212EA 312.97 USD
76981-212 76981-214 76981-216
4-Chloro-6-methyl-5H-pyrrolo[3,2-d]pyrimidine 95%
4-Chloro-6-methyl-5H-pyrrolo[3,2-d]pyrimidine

Precaución: for Research Use Only

Formula: C₇H₆ClN₃
MW: 167.59 g/mol
MDL Number: MFCD17013038
CAS Number: 91862-35-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR