tert-Butyl (3-hydroxybutyl)carbamate 97%

Supplier: Ambeed
Warning

A391056-250MG A391056-1G
76978-944EA 147 USD
76978-944 77882-043
tert-Butyl (3-hydroxybutyl)carbamate 97%
tert-Butyl (3-hydroxybutyl)carbamate

Precaución: for Research Use Only

Formula: C₉H₁₉NO₃
MW: 189.25 g/mol
MDL Number: MFCD04974277
CAS Number: 167216-30-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR