5-Formyl-2-methoxyphenyl Acetate 97%

Supplier: Ambeed
Warning

Synonyms: Acetic acid 5-Formyl-2-methoxyphenyl ester, 3-Acetoxy-4-methoxybenzaldehyde

A798591-1G A798591-5G A798591-25G
76981-658EA 10 USD
76981-658 76981-660 77882-719
5-Formyl-2-methoxyphenyl Acetate 97%
5-Formyl-2-methoxyphenyl Acetate

Precaución: for Research Use Only

Formula: C₁₀H₁₀O₄
MDL Number: MFCD00266441
CAS Number: 881-57-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR