7-Bromo-4-chloro-3-nitroquinoline 95%

Supplier: Ambeed
Warning

A145708-1G A145708-250MG
76972-828EA 206.88 USD
76972-828 76972-830
7-Bromo-4-chloro-3-nitroquinoline 95%
7-Bromo-4-chloro-3-nitroquinoline

Precaución: for Research Use Only

Formula: C₉H₄BrClN₂O₂
MW: 287.5000 g/mol
MDL Number: MFCD12828220
CAS Number: 723280-98-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR