2-(1H-Benzimidazol-2-yl)ethanamine dihydrochloride ≥99%

Supplier: Ambeed
Warning

Synonyms: 2-(2-Aminoethyl)benzimidazole dihydrochloride, 2-(1H-Benzo[d]imidazol-2-yl)ethanamine dihydrochloride

A377466-100MG A377466-10G A377466-1G A377466-250MG A377466-25G A377466-5G
76978-796EA 5 USD
76978-796 76978-798 76978-800 76978-802 76978-804 77882-013
2-(1H-Benzimidazol-2-yl)ethanamine dihydrochloride ≥99%
2-(1H-Benzimidazol-2-yl)ethanamine dihydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₃Cl₂N₃
MDL Number: MFCD01325081
CAS Number: 4499-07-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR