3-Amino-N-methylbenzenesulfonamide 97%

Supplier: Ambeed
Warning

A478908-1G A478908-250MG A478908-5G
76979-726EA 90.75 USD
76979-726 76979-728 76979-730
3-Amino-N-methylbenzenesulfonamide 97%
3-Amino-N-methylbenzenesulphonamide

Precaución: for Research Use Only

Formula: C₇H₁₀N₂O₂S
MDL Number: MFCD07363815
CAS Number: 459434-40-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR