1-Methyl-3-(trifluoromethyl)-1H-pyrazol-4-amine 97%

Supplier: Ambeed
Warning

A298167-100MG A298167-1G A298167-250MG
76977-678EA 15 USD
76977-678 76977-680 76977-682
1-Methyl-3-(trifluoromethyl)-1H-pyrazol-4-amine 97%
1-Methyl-3-(trifluoromethyl)-1H-pyrazol-4-amine

Precaución: for Research Use Only

Formula: C₅H₆F₃N₃
MDL Number: MFCD04970058
CAS Number: 1006436-44-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR