Methyl 3-cyano-4-hydroxybenzoate 98%
Supplier: Ambeed
|
Warning
|
A180544-100MG
A180544-1G
A180544-250MG
76974-444EA
60.48
USD
76974-444
76974-446
76974-448
Methyl 3-cyano-4-hydroxybenzoate 98%
Methyl 3-cyano-4-hydroxybenzoate
Precaución: for Research Use Only
Formula:
C₉H₇NO₃ MW: 177.157 g/mol |
MDL Number:
MFCD13248596 CAS Number: 156001-68-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...