C-(1-Methylpyrrolidin-3-yl)methylamine 97%

Supplier: Ambeed
Danger

Synonyms: (1-Methylpyrrolidin-3-yl)methanamine

A495973-100MG A495973-1G A495973-250MG A495973-5G
76979-818EA 56.23 USD
76979-818 76979-820 76979-822 76979-824
C-(1-Methylpyrrolidin-3-yl)methylamine 97%
C-(1-Methylpyrrolidin-3-yl)methylamine

Precaución: for Research Use Only

Formula: C₆H₁₄N₂
MW: 114.19 g/mol
MDL Number: MFCD05864515
CAS Number: 13005-11-3
UN: 2735
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR