3-Bromo-2-chloroanisole 98%

Supplier: Ambeed
Warning

Synonyms: 1-Bromo-2-chloro-3-methoxybenzene

A637133-1G A637133-250MG
76980-722EA 46.68 USD
76980-722 76980-724
3-Bromo-2-chloroanisole 98%
3-Bromo-2-chloroanisole

Precaución: for Research Use Only

Formula: C₇H₆BrClO
MW: 221.4789 g/mol
MDL Number: MFCD16036127
CAS Number: 174913-11-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR