(1-Ethyl-4-piperidinyl)methanol 95%

Supplier: Ambeed
Warning

Synonyms: (1-Ethylpiperidin-4-yl)methanol

A572176-1G A572176-5G A572176-25G
76980-344EA 36 USD
76980-344 77673-540 77882-370
(1-Ethyl-4-piperidinyl)methanol 95%
(1-Ethyl-4-piperidinyl)methanol

Precaución: for Research Use Only

Formula: C₈H₁₇NO
MDL Number: MFCD08059823
CAS Number: 90226-87-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR