3-Bromo-5-(methylsulfonyl)pyridine 95%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-3-methylsulphonylpyridine, 5-Bromopyridin-3-yl methyl sulphone

A781142-1G A781142-250MG
76981-578EA 109.28 USD
76981-578 76981-580
3-Bromo-5-(methylsulfonyl)pyridine 95%
3-Bromo-5-(methylsulfonyl)pyridine

Precaución: for Research Use Only

Formula: C₆H₆BrNO₂S
MW: 236.086 g/mol
MDL Number: MFCD09801057
CAS Number: 445491-71-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR