3-Fluoro-2-methoxybenzyl alcohol 95%

Supplier: Ambeed
Warning

Synonyms: 3-Fluoro-2-methoxybenzenemethanol

A127035-10G A127035-1G
76971-904EA 210 USD
76971-904 76971-906
3-Fluoro-2-methoxybenzyl alcohol 95%
3-Fluoro-2-methoxybenzyl alcohol

Precaución: for Research Use Only

Formula: C₈H₉FO₂
MDL Number: MFCD09025369
CAS Number: 303043-91-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR