2-Methoxy-3-(trifluoromethyl)-5-pyridineboronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: 2-Methoxy-3-(trifluoromethyl)pyridine-5-boronic Acid Pinacol Ester

A481036-100MG A481036-1G A481036-250MG
76979-748EA 35 USD
76979-748 76979-750 76979-752
2-Methoxy-3-(trifluoromethyl)-5-pyridineboronic acid pinacol ester 98%
2-Methoxy-3-(trifluoromethyl)-5-pyridineboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₇BF₃NO₃
MW: 303.0900 g/mol
MDL Number: MFCD12026094
CAS Number: 1150561-61-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR