tert-butyl 1-benzyl-3-(hydroxymethyl)pyrrolidin-3-ylcarbamate 95%

Supplier: Ambeed
Warning

A888460-100MG A888460-250MG
76981-886EA 96.55 USD
76981-886 76981-888
tert-butyl 1-benzyl-3-(hydroxymethyl)pyrrolidin-3-ylcarbamate 95%
tert-butyl 1-benzyl-3-(hydroxymethyl)pyrrolidin-3-ylcarbamate

Precaución: for Research Use Only

Formula: C₁₇H₂₆N₂O₃
MW: 306.39 g/mol
MDL Number: MFCD06410561
CAS Number: 475469-14-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR