Methyl-2-bromo-4-chlorobenzoate 98%

Supplier: Ambeed
Warning

Synonyms: 2-Bromo-4-chlorobenzoic acid methyl ester, Methyl 2-bromo-4-chlorobenzoate

A194109-10G A194109-1G A194109-25G A194109-5G
76974-970EA 35.01 USD
76974-970 76974-972 76974-974 76974-976
Methyl-2-bromo-4-chlorobenzoate 98%
Methyl-2-bromo-4-chlorobenzoate

Precaución: for Research Use Only

Formula: C₈H₆BrClO₂
MW: 249.4900 g/mol
MDL Number: MFCD10566822
CAS Number: 57381-62-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR