2-Bromo-6-(2-methyl-1,3-dioxolan-2-yl)pyridine 97%

Supplier: Ambeed
Warning

A206638-10G A206638-1G A206638-250MG A206638-25G A206638-5G
76975-370EA 93.36 USD
76975-370 76975-372 76975-374 76975-376 76975-378
2-Bromo-6-(2-methyl-1,3-dioxolan-2-yl)pyridine 97%
2-Bromo-6-(2-methyl-1,3-dioxolan-2-yl)pyridine

Precaución: for Research Use Only

Formula: C₉H₁₀BrNO₂
MW: 244.09 g/mol
MDL Number: MFCD12406888
CAS Number: 49669-14-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR