1-(2-Amino-5-chlorophenyl)-2,2,2-trifluoroethanone hydrochloride hydrate 98%
Supplier: Ambeed
|
Warning
|
A450822-1G
A450822-25G
A450822-5G
76979-488EA
11
USD
76979-488
76979-490
76979-492
1-(2-Amino-5-chlorophenyl)-2,2,2-trifluoroethanone hydrochloride hydrate 98%
1-(2-Amino-5-chlorophenyl)-2,2,2-trifluoroethanone hydrochloride hydrate
Precaución: for Research Use Only
Formula:
C₈H₈Cl₂F₃NO₂ |
MDL Number:
MFCD09841379 CAS Number: 1184936-21-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...