D-Tryptophanol 98%

Supplier: Ambeed
Warning

Synonyms: (R)-2-Amino-3-(1H-indol-3-yl)propan-1-ol

A239541-1G A239541-250MG A239541-5G
76976-404EA 45.62 USD
76976-404 76976-406 76976-408
D-Tryptophanol 98%
D-Tryptophanol

Precaución: for Research Use Only

Formula: C₁₁H₁₄N₂O
MW: 190.246 g/mol
MDL Number: MFCD00270220
CAS Number: 52485-52-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR