N(α)-Boc-D-tryptophan 98%

Supplier: Ambeed
Warning

A280171-100G A280171-10G A280171-1G A280171-25G A280171-500G A280171-5G
76977-386EA 44 USD
76977-386 76977-388 76977-390 76977-392 76977-394 76977-396
N(α)-Boc-D-tryptophan 98%
N(α)-Boc-D-tryptophan

Precaución: for Research Use Only

Formula: C₁₆H₂₀N₂O₄
MDL Number: MFCD00037944
CAS Number: 5241-64-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR