Dimenhydrinate 98%

Supplier: Ambeed
Warning

Synonyms: Amosyt, 8-Chloro-1,3-dimethyl-1H-purine-2,6(3H,7H)-dione 2-(benzhydryloxy)-N,N-dimethylethanamine salt

A814705-100MG A814705-1G
76981-706EA 5.31 USD
76981-706 76981-708
Dimenhydrinate 98%
Dimenhydrinate

Precaución: for Research Use Only

Formula: C₁₇H₂₁NO·C₇H₇ClN₄O₂
MW: 469.96 g/mol
MDL Number: MFCD00054265
CAS Number: 523-87-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR