1-Phenyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole 97%

Supplier: Ambeed
Warning

Synonyms: 1-Phenylpyrazole-4-boronic acid, pinacol ester, 1-Phenyl-1H-pyrazole-4-boronic acid pinacol ester

A262241-100MG A262241-250MG A262241-5G A262241-1G
76976-982EA 18 USD
76976-982 76976-984 77672-348 77672-346
1-Phenyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole 97%
1-Phenyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole

Precaución: for Research Use Only

Formula: C₁₅H₁₉BN₂O₂
MDL Number: MFCD12400941
CAS Number: 1002334-12-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR