5-Methyl-1H-indazole-4-boronic acid 97%

Supplier: Ambeed
Warning

Synonyms: (5-Methyl-1H-indazol-4-yl)boronic acid

A332586-100MG A332586-10G A332586-1G A332586-250MG A332586-25G A332586-5G
76978-272EA 5 USD
76978-272 76978-274 76978-276 76978-278 76978-280 76978-282
5-Methyl-1H-indazole-4-boronic acid 97%
5-Methyl-1H-indazole-4-boronic acid

Precaución: for Research Use Only

Formula: C₈H₉BN₂O₂
MDL Number: MFCD10001490
CAS Number: 1245816-10-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR