1-Ethyl-3-methylimidazolium bromide 97%

Supplier: Ambeed
Warning

A250140-25G A250140-100G A250140-500G
76976-668EA 11 USD
76976-668 77881-665 77881-666
1-Ethyl-3-methylimidazolium bromide 97%
1-Ethyl-3-methylimidazolium bromide

Precaución: for Research Use Only

Formula: C₆H₁₁BrN₂
MDL Number: MFCD03427610
CAS Number: 65039-08-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR