(1R,2R)-(-)-N,N'-Dimethylcyclohexane-1,2-diamine 97%

Supplier: Ambeed
Danger

Synonyms: (1R,2R)-(-)-1,2-Bis(methylamino)cyclohexane, (1R,2R)-N,N'-Dimethyl-1,2-cyclohexanediamine

A383233-10G A383233-1G A383233-25G A383233-5G
76978-846EA 21 USD
76978-846 76978-848 77551-116 77551-118
(1R,2R)-(-)-N,N'-Dimethylcyclohexane-1,2-diamine 97%
(1R,2R)-(-)-N,N'-Dimethylcyclohexane-1,2-diamine

Precaución: for Research Use Only

Formula: C₈H₁₈N₂
MW: 142.25 g/mol
MDL Number: MFCD00671527
CAS Number: 68737-65-5
UN: 3259
ADR: 8 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR