Ethyl-2-bromo-4H-thieno[3,2-b]pyrrole-5-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: Ethyl 2-bromo-4H-thieno[3,2-b]pyrrole-5-carboxylate

A229365-1G A229365-5G
76976-088EA 164.44 USD
76976-088 76976-090
Ethyl-2-bromo-4H-thieno[3,2-b]pyrrole-5-carboxylate 95%
Ethyl-2-bromo-4H-thieno[3,2-b]pyrrole-5-carboxylate

Precaución: for Research Use Only

Formula: C₉H₈BrNO₂S
MW: 274.132 g/mol
MDL Number: MFCD15730638
CAS Number: 238749-50-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR