4-Bromo-3-fluoropyridine 98%
Supplier: Ambeed
|
Warning
|
A427048-1G
A427048-250MG
A427048-5G
A427048-25G
76979-292EA
28.99
USD
76979-292
76979-294
76979-296
77882-116
4-Bromo-3-fluoropyridine 98%
4-Bromo-3-fluoropyridine
Precaución: for Research Use Only
Formula:
C₅H₃BrFN |
MDL Number:
MFCD04113266 CAS Number: 2546-52-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...